EMAIL THIS PAGE TO A FRIEND
Cell
Small talk. Cell-to-cell communication in bacteria.
PMID 12086599
Abstract
In a process called quorum sensing, groups of bacteria communicate with one another to coordinate their behavior and function like a multicellular organism. A diverse array of secreted chemical signal molecules and signal detection apparatuses facilitate highly productive intra- and interspecies relationships.