EMAIL THIS PAGE TO A FRIEND
Planta medica
Inhibitory effects of natural sesquiterpenoids isolated from the rhizomes of Curcuma zedoaria on prostaglandin E2 and nitric oxide production.
PMID 12094302
Abstract
Beta-turmerone and ar-turmerone, sesquiterpenoids isolated from the rhizome of Curcuma zedoaria, inhibited lipopolysaccharide (LPS)-induced prostaglandin E 2 production in cultured mouse macrophage cell RAW 264.7 in a dose-dependent manner (IC 50 = 7.3 microM for beta-turmerone; IC 50 = 24.0 microM for ar-turmerone). In addition, these compounds exhibited inhibitory effects on LPS-induced nitric oxide production in the cell system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42258
(S)-ar-Turmerone, analytical standard
C15H20O