EMAIL THIS PAGE TO A FRIEND
Methods (San Diego, Calif.)
Measurement of genome wide DNA methylation by reversed-phase high-performance liquid chromatography.
PMID 12095275
Abstract
A method to determine the extent of cytosine methylation in DNA is described. The technique involves the enzymatic hydrolysis of DNA to its deoxyribonucleotide components and subsequent separation and quantification of the nucleotides by isocratic reversed-phase high-performance liquid chromatography (HPLC). The system gives highly reproducible results and, under suitable conditions, is capable of measuring 5-methylcytosine levels in as little as 1 microg of DNA.