EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Histone 2A stimulates glucose-6-phosphatase activity by permeabilization of liver microsomes.
PMID 12097138
Abstract
Histone 2A increases glucose-6-phosphatase activity in liver microsomes. The effect has been attributed either to the conformational change of the enzyme, or to the permeabilization of microsomal membrane that allows the free access of substrate to the intraluminal glucose-6-phosphatase catalytic site. The aim of the present study was the critical reinvestigation of the mechanism of action of histone 2A. It has been found that the dose-effect curve of histone 2A is different from that of detergents and resembles that of the pore-forming alamethicin. Inhibitory effects of EGTA on glucose-6-phosphatase activity previously reported in histone 2A-treated microsomes have been also found in alamethicin-permeabilized vesicles. The effect of EGTA cannot therefore simply be an antagonization of the effect of histone 2A. Histone 2A stimulates the activity of another latent microsomal enzyme, UDP-glucuronosyltransferase, which has an intraluminal catalytic site. Finally, histone 2A renders microsomal vesicles permeable to non-permeant compounds. Taken together, the results demonstrate that histone 2A stimulates glucose-6-phosphatase activity by permeabilizing the microsomal membrane.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
67328
Methyl 6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside, ≥97.0% (TLC)
C15H29NO7