EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Accurate mass analysis of ethanesulfonic acid degradates of acetochlor and alachlor using high-performance liquid chromatography and time-of-flight mass spectrometry.
PMID 12102310
Abstract
Degradates of acetochlor and alachlor (ethanesulfonic acids, ESAs) were analyzed in both standards and in a groundwater sample using high-performance liquid chromatography-time-of-flight mass spectrometry with electrospray ionization. The negative pseudomolecular ion of the secondary amide of acetochlor ESA and alachlor ESA gave average masses of 256.0750+/-0.0049 amu and 270.0786+/-0.0064 amu respectively. Acetochlor and alachlor ESA gave similar masses of 314.1098+/-0.0061 amu and 314.1153+/-0.0048 amu; however, they could not be distinguished by accurate mass because they have the same empirical formula. On the other hand, they may be distinguished using positive-ion electrospray because of different fragmentation spectra, which did not occur using negative-ion electrospray.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471550
Ethanesulfonic acid solution, 70 wt. % in H2O
C2H6O3S