EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Diastereoselective formation of glycoluril dimers: isomerization mechanism and implications for cucurbit[n]uril synthesis.
PMID 12105910
Abstract
Cucurbit[6]uril (CB[6]) is a macrocyclic compound, prepared in one pot from glycoluril and formaldehyde, whose molecular recognition properties have made it the object of intense study. Studies of the mechanism of CB[n] formation, which might provide insights that allow the tailor-made synthesis of CB[n] homologues and derivatives, have been hampered by the complex structure of CB[n]. By reducing the complexity of the reaction to the formation of S-shaped (12S-18S) and C-shaped (12C-18C) methylene bridged glycoluril dimers, we have been able to probe the fundamental steps of the mechanism of CB[n] synthesis to a level that has not been possible previously. For example, we present strong evidence that the mechanism of CB[n] synthesis proceeds via the intermediacy of both S-shaped and C-shaped dimers. The first experimental determination of the relative free energies of the S-shaped and C-shaped dimers indicates a thermodynamic preference (1.55-3.25 kcal mol(-)(1)) for the C-shaped diastereomer. This thermodynamic preference is not because of self-association, solvation, or template effects. Furthermore, labeling experiments have allowed us to elucidate the mechanism of this acid-catalyzed equilibrium between the S-shaped and C-shaped diastereomers. The equilibration is an intramolecular process that proceeds with high diastereoselectivity and retention of configuration. On the basis of the broad implications of these results for CB[n] synthesis, we suggest new synthetic strategies that may allow for the improved preparation of CB[n] (n > 8) and CB[n] derivatives from functionalized glycolurils.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G7305
Glycoluril, 97%
C4H6N4O2