EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
K+-induced hyperpolarization in rat mesenteric artery: identification, localization and role of Na+/K+-ATPases.
PMID 12110616
Abstract
1. Mechanisms underlying K(+)-induced hyperpolarizations in the presence and absence of phenylephrine were investigated in endothelium-denuded rat mesenteric arteries (for all mean values, n=4). 2. Myocyte resting membrane potential (m.p.) was -58.8+/-0.8 mV. Application of 5 mM KCl produced similar hyperpolarizations in the absence (17.6+/-0.7 mV) or presence (15.8+/-1.0 mV) of 500 nM ouabain. In the presence of ouabain +30 microM barium, hyperpolarization to 5 mM KCl was essentially abolished. 3. In the presence of 10 microM phenylephrine (m.p. -33.7+/-3 mV), repolarization to 5 mM KCl did not occur in the presence or absence of 4-aminopyridine but was restored (-26.9+/-1.8 mV) on addition of iberiotoxin (100 nM). Under these conditions the K+-induced repolarization was insensitive to barium (30 microM) but abolished by 500 nM ouabain alone. 4. In the presence of phenylephrine + iberiotoxin the hyperpolarization to 5 mM K(+) was inhibited in the additional presence of 300 nM levcromakalim, an action which was reversed by 10 microM glibenclamide. 5. RT-PCR, Western blotting and immunohistochemical techniques collectively showed the presence of alpha(1)-, alpha(2)- and alpha(3)-subunits of Na(+)/K(+)-ATPase in the myocytes. 6. In K(+)-free solution, re-introduction of K(+) (to 4.6 mM) hyperpolarized myocytes by 20.9+/-0.5 mV, an effect unchanged by 500 nM ouabain but abolished by 500 microM ouabain. 7. We conclude that under basal conditions, Na(+)/K(+)-ATPases containing alpha(2)- and/or alpha(3)-subunits are partially responsible for the observed K(+)-induced effects. The opening of myocyte K(+) channels (by levcromakalim or phenylephrine) creates a 'K(+) cloud' around the cells which fully activates Na(+)/K(+)-ATPase and thereby abolishes further responses to [K(+)](o) elevation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2714 Protease Inhibitor Cocktail powder, for general use, lyophilized powder