EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Lung Cellular and Molecular Physiology
Enhanced nitric oxide and reactive oxygen species production and damage after inhalation of silica.
PMID 12114212