EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical sciences
Theoretical and experimental investigations on the morphology of pharmaceutical crystals.
PMID 12115826
Abstract
We investigated the morphologies of three polymorphs of 1,3-di(cyclopropylmethyl)-8-aminoxanthine, a compound of pharmaceutical importance. We compared the experimental morphologies with those predicted by theoretical methods. We also predicted the elastic constants of the three polymorphs. These results are used to help assess the stabilities of the three polymorphs and compare the abilities of theoretical methods to reproduce experimental morphologies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0366
Cipamfylline, ≥98% (HPLC)
C13H17N5O2