EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Enantioseparation of glutethimide and its 5-OH-metabolite in capillary electrophoresis and study of selector-selectand interactions using one-dimensional rotating frame nuclear Overhauser and exchange spectroscopy.
PMID 12116135
Abstract
Enantioseparation of glutethimide (GT) and its 5-hydroxy metabolite (5-OH-GT) has been studied with several charged cyclodextrin (CD) derivatives. The emphasis was made on the enantiomer migration order of GT and simultaneous enantioseparation of GT and 5-OH-GT. The possible structural differences of GT complexes with three different single isomer charged CD derivatives were studied using one-dimensional rotating frame nuclear Overhauser and exchange spectroscopy (1-D ROESY).
Related Materials