EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Tandem intermolecular Suzuki coupling/intramolecular vinyl triflate-arene coupling.
PMID 12123001
Abstract
Treatment of a benzyl substituted meso-ditriflate with boronic acids in the presence of palladium acetate, triphenylphosphine and caesium fluoride results in intermolecular Suzuki coupling followed by vinyl triflate-arene cyclisation to provide, in high yields, single regioisomers of tricyclic-carbocycles.