EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
The structures of Cu(I) and Ag(I) coordination polymers using the tricyanofluoroborate anion.
PMID 12123006
Abstract
The anion BF(CN)3-forms isomorphous network polymers with Cu(I) and Ag(I) that exhibit one-dimensional channels along the b axis and demonstrate stability to air and light respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
207934
Tetrafluoroboric acid solution, 48 wt. % in H2O
HBF4