EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
TS-1 zeolite microengineered reactors for 1-pentene epoxidation.
PMID 12123024
Abstract
A zeolite-based microengineered reactor was fabricated and tested for 1-pentene epoxidation over titanium silicalite-1 (TS-1) catalyst, which has been selectively incorporated within the microreactor channel using a new synthesis procedure.
Related Materials