EMAIL THIS PAGE TO A FRIEND
Organic letters
First asymmetric total synthesis of (+)-sparteine.
PMID 12123380
Abstract
[reaction: see text] The total synthesis of (+)-sparteine was accomplished from 2,5-norbornadione in 15 steps and 15.7% overall yield. The key steps were two ring-expansion reactions, one involving an intramolecular Schmidt reaction and one using a novel variant of the photo-Beckmann rearrangement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B33803
Bicyclo[2.2.1]hepta-2,5-diene, 98%
C7H8