EMAIL THIS PAGE TO A FRIEND
FEBS letters
Identification of prolylcarboxypeptidase as the cell matrix-associated prekallikrein activator.
PMID 12123826
Abstract
Investigations determined that the cell matrix-associated prekallikrein (PK) activator is prolylcarboxypeptidase. PK activation on human umbilical vein endothelial cell (HUVEC) matrix is inhibited by antipain (IC(50)=50 microM) but not anti-factor XIIa antibody, 3 mM benzamidine, 5 mM iodoacetic acid or iodoacetamide, or 3 mM N-ethylmaleimide. Corn trypsin inhibitor (IC(50)=100 nM) or Fmoc-aminoacylpyrrolidine-2-nitrile (IC(50)=100 microM) blocks matrix-associated PK activation. Angiotensin II (IC(50)=100 microM) or bradykinin (IC(50)=3 mM), but not angiotensin 1-7 or bradykinin 1-5, inhibits matrix-associated PK activation. ECV304 cell matrix PK activator also is blocked by 100 microM angiotensin II, 1 microM corn trypsin inhibitor, and 50 microM antipain, but not angiotensin 1-7. 1 mM angiotensin II or 300 microM Fmoc-aminoacylpyrrolidine-2-nitrile indirectly blocks plasminogen activation by inhibiting kallikrein formation for single chain urokinase activation. On immunoblot, prolylcarboxypeptidase antigen is associated with HUVEC matrix. These studies indicate that prolylcarboxypeptidase is the matrix PK activator.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
10791
Antipain, >50000 U/mg
C27H44N10O6