EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Peroxisomal fatty acid oxidation and cellular redox.
PMID 12125363
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1514
Nervonic acid, ≥99% (capillary GC)
C24H46O2
87117
Nervonic acid, analytical standard
C24H46O2