EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
An improved protocol for the preparation of 3-pyridyl- and some arylboronic acids.
PMID 12126436
Abstract
3-Pyridylboronic acid was prepared in high yield and bulk quantity from 3-bromopyridine via a protocol of lithium-halogen exchange and "in situ quench". This technique was further studied and evaluated on other aryl halides in the preparation of arylboronic acids.