EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
High-resolution separation of polychlorinated biphenyls by comprehensive two-dimensional gas chromatography.
PMID 12134818
Abstract
Comprehensive two-dimensional gas chromatography (GC x GC) with micro electron-capture detection (microECD) has been optimised for the separation of polychlorinated biphenyl congeners with emphasis on the separation of 12 toxic non- and mono-ortho chlorinated biphenyls (CBs), viz. CBs 77, 81, 105, 114, 118, 123, 126, 156, 157, 167, 169 and 189. The selection of the first- and second-dimension columns and the temperature programme optimisation were carried out with a mixture of 90 CBs and the results are compared with those of one-dimensional GC. A complete separation of all 12 priority CBs was obtained with two column combinations, HP-1-HT-8 and HP-1-SupelcoWax-10. With the HP-1-HT-8 column set, ordered structures show up in the two-dimensional plane, with the number of chlorine substituents and their position (ortho vs. non-ortho) being the main parameters of interest. This can help with congener identification. Estimated detection limits are excellent, i.e. about 10 fg. To illustrate the potential and the versatility of GC x GC-microECD, a cod liver extract and a standard mixture of the 17 most toxic polychlorinated dibenzo-p-dioxins and polychlorinated dibenzofurans together with 90 CBs were analysed as an application.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
25026-U
SUPELCOWAX® 10 Capillary GC Column, L × I.D. 10 m × 0.10 mm, df 0.10 μm
24343 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm
24169 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.20 mm, df 0.20 μm
24170 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.20 mm, df 0.20 μm
24077 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
24079 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
24081 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
23308-U SUPELCOWAX® 10 Capillary GC Column, df 0.25 μm
24284 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
24078 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
24080-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
24082 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
24084 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.50 μm
24085-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.50 μm
24211 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 1.00 μm
24212 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.00 μm
25325 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 0.50 μm
25385 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 0.50 μm
25300-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 1.00 μm
25301-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 1.00 μm
25391 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 1.00 μm
25375-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 2.00 μm
25376 SUPELCOWAX® 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 2.00 μm
23327-U SUPELCOWAX® 10 Capillary GC Column, L × I.D. 30 m × 0.75 mm, df 1.00 μm