EMAIL THIS PAGE TO A FRIEND
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova
[Effects of natural (zinc, vitamin E) and synthetic (diludin) antioxidants on the intestinal permeability in chicks with vitamin A deficiency].
PMID 12136734
Abstract
The main concern of this work was to examine the relation between altered antioxidant status on the one hand and increase in L-tryptophan absorption in the small intestine in order to bring further information regarding to possible role of vitamin A and zinc to maintaining of intestinal epithelial barrier integrity, on the other hand. In control, only some ideal tight junctions at the tip of the villi were permeable to ZnC1(2), whereas in A-hypovitaminosis permeability increased significantly. Studies demonstrate that an increased L-tryptophan accumulation in the intestinal mucous may result from a free radical damage to the mucous surface with formation of "leaky" junctions in the ilea. The results suggest that the zinc plays a crucial role in stabilizing biomembranes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
120227
Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, 95%
C13H19NO4