EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Drug-induced translocation of nucleolar proteins fused to green fluorescent protein.
PMID 12136750
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3580
Toyocamycin, ≥98% (HPLC), from Streptomyces rimosus
C12H13N5O4