EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Mitochondrial aspartate aminotransferase catalyses cysteine S-conjugate beta-lyase reactions.
PMID 12137566
Abstract
Rat liver mitochondrial aspartate aminotransferase (a homodimer) was shown to catalyse a beta-lyase reaction with three nephrotoxic halogenated cysteine S-conjugates [ S -(1,1,2,2-tetrafluoroethyl)-L-cysteine, S -(1,2-dichlorovinyl)-L-cysteine and S -(2-chloro-1,1,2-trifluoroethyl)-L-cysteine], and less effectively so with a non-toxic cysteine S-conjugate [benzothiazolyl-L-cysteine]. Transamination competes with the beta-lyase reaction, but is not favourable. The ratio of beta elimination to transamination in the presence of S -(1,1,2,2-tetrafluoroethyl)-L-cysteine and 2-oxoglutarate is >100. Syncatalytic inactivation by the halogenated cysteine S-conjugates is also observed. The enzyme turns over approx. 2700 molecules of halogenated cysteine S-conjugate on average for every monomer inactivated. Kidney mitochondria are known to be especially sensitive to toxic halogenated cysteine S-conjugates. Evidence is presented that 15-20% of the cysteine S-conjugate beta-lyase activity towards S -(1,1,2,2-tetrafluoroethyl)-L-cysteine in crude kidney mitochondrial homogenates is due to mitochondrial aspartate aminotransferase. The possible involvement of mitochondrial aspartate aminotransferase in the toxicity of halogenated cysteine S-conjugates is also discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4284
β-Chloro-D-alanine hydrochloride
C3H6ClNO2 · HCl
C9033
β-Chloro-L-alanine hydrochloride, Alanine aminotransferase inhibitor
C3H6ClNO2 · HCl