EMAIL THIS PAGE TO A FRIEND
Sheng wu hua xue yu sheng wu wu li xue bao Acta biochimica et biophysica Sinica
Expression of Brain-derived Neurotrophic Factor (BDNF) Gene in Rat Myoblast Cells.
PMID 12142916
Abstract
Brain-derived neurotrophic factor (BDNF) gene was cloned into retrovirous vector plasmid pLNCX to form the pLNC/BDNF. After packaging in PA317 cell line, the infectious particles were used to infect rat myoblast cell line L-6TG. After selection with G418, the L-6TG/BDNF cells were collected. Results from Southern blot show that bdnf gene has successfully integrated into the L-6TG genomic DNA. The expression of bdnf gene has been proved by Northern blot and Dot blot. The content of BDNF in the culture medium of L-6TG/BDNF was about 25 ng.10(-6) cells per 24 h per ml.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
PA317, 89032007