EMAIL THIS PAGE TO A FRIEND
Cryobiology
Thermal study of simple amino-alcohol solutions.
PMID 12151270
Abstract
Widely regarded as the most promising approach to long-term cryopreservation of organs for transplantation, vitrification is a process where liquid is transformed into a disordered solid state free from crystals, known as the amorphous state. The vitreous state is obtained by rapid cooling to cryogenic temperatures in the presence of antifreeze substances called cryoprotectants, such as polyalcohols, which are known to be very good vitrification agents. This work reports on the thermal properties of a new class of compounds, the amino-alcohols, studied for its similarity to the structure of the equivalent polyalcohols. We studied by differential scanning calorimetry the glass-forming tendency and stability of the amorphous state for de-ionized water solutions containing 2-amino-1-ethanol and 3-amino-1-propanol at the concentrations of 35%, 40%, 43%, and 45% (w/w). A comparison is made with previous results obtained by Mehl [Cryobiology 27 (1990) 687-688] on the same compounds under different experimental conditions. The results are also compared with those obtained by Boutron [Cryobiology 30 (1993) 87-97] for the corresponding dialcohols. A further comparison is made with a few results obtained for the 1-amino-2-propanol and the 2-amino-1-propanol tested under the same conditions.