EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Cytofluorescence techniques for the visualization of distinct pools of protein thiols at the single cell level.
PMID 12152209
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94506
Tetramethylrhodamine-5-maleimide, suitable for fluorescence, ≥85% (HPLC)
C28H23N3O5