EMAIL THIS PAGE TO A FRIEND
Organic letters
Effect of ligand structure on the zinc-catalyzed Henry reaction. Asymmetric syntheses of (-)-denopamine and (-)-arbutamine.
PMID 12153193
Abstract
[reaction: see text] Syntheses of variously modified ligands for the dinuclear zinc catalysts for the asymmetric aldol and nitroaldol (Henry) reactions are reported. Catalytic enantioselective nitroaldol reactions promoted by these modified ligands led to efficient syntheses of the beta-receptor agonists (-)-denopamine and (-)-arbutamine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7815
R(−)-Denopamine, ≥98% (HPLC), powder
C18H23NO4