EMAIL THIS PAGE TO A FRIEND
Organic letters
A one-pot access to 6-substituted phenanthridines from fluoroarenes and nitriles via 1,2-arynes.
PMID 12153210
Abstract
[reaction: see text] A one-pot, t-BuLi-induced synthesis of 6-substituted phenanthridines from fluoroarenes and nitriles via 1,2-arynes is reported. Aryl- and hetaryl nitriles, cyanamides, and trimethylacetonitrile gave phenanthridine products. The method was extended to provide bisphenanthridine 10 by a one-pot bis-cyclization, using 1,3-dicyanobenzene and PhF in 1:5 ratio. Reaction of 1-fluoronaphthalene and 4-chlorofluorobenzene with benzonitrile afforded the regioisomerically pure products 11 and 12, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196657
1-Fluoronaphthalene, 99%
C10H7F