EMAIL THIS PAGE TO A FRIEND
Preparative biochemistry
A large scale procedure for purification of desmosine and isodesmosine.
PMID 1215339
Abstract
A rapid large scale procedure was devised for the purification of desmosine and isodesmosine from ligamentum nuchae elastin. The method makes use of the hydrophilic nature of the desmosines which preferentially absorbs to cellulose fibers in mixtures of organic solvents. Resolution of the isomers was achieved on a polystyrene resin column.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9439
Desmosine chloride, from bovine, 99.5%±0.2%
C24H40ClN5O8