EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Studies of the constituents of Uruguayan propolis.
PMID 12166959
Abstract
Eighteen flavonoids including two new compounds, four aromatic carboxylic acids, and eleven phenolic acid esters including one new compound were isolated and identified from the ethyl acetate soluble fraction of the 70% ethanol extract of Uruguayan propolis. The new compounds were elucidated as pinobanksin 3-(2-methyl)butyrate (1; recently reported in Usia, T.; Banskota, A. H.; Tezuka, Y.; Midorikawa, K.; Matsushige, K.; Kadota, S. J. Nat. Prod. 2002, 65, 673-676) pinobanksin 3-isobutyrate (2), and 2-methyl-2-butenyl ferulate (24). The constituents isolated from Uruguayan propolis in this study were similar to those of propolis of European and Chinese origin. Thus, it is suggested that the Uruguayan propolis has a plant origin similar to those of propolis from Europe and China.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68530
Pinobanksin, ≥95% (HPLC)
C15H12O5