EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
New sources of dietary myosmine uptake from cereals, fruits, vegetables, and milk.
PMID 12166981
Abstract
Myosmine has been regarded as a specific tobacco alkaloid until investigations pointed out that nuts and nut products constitute a significant source of myosmine. In the present study it is shown that the occurrence of myosmine is widespread throughout a large number of plant families. Using a method for extraction practicable for all examined foods, quantitative analysis through internal standard addition showed nanograms per gram amounts. Positively tested edibles were staple foods such as maize, rice, wheat flour, millet, potato, and milk and also cocoa, popcorn, tomato, carrot, pineapple, kiwi, and apples. No myosmine was detectable in other vegetables and fruits such as lettuce, spinach, cucumber, onion, banana, tangerines, and grapes. Myosmine is easily nitrosated giving rise to a DNA adduct identical to the esophageal tobacco carcinogen N-nitrosonornicotine. Therefore, the role of dietary myosmine in esophageal adenocarcinoma should be further investigated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8765
Myosmine, ≥98%
C9H10N2