EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Isoserine-based biotinylated photoaffinity probes that interact with penicillin-binding protein 1b.
PMID 12170816
Abstract
The photolytic decomposition of trifunctional carbene generating photoaffinity probes in methanolic solution was studied, a cleavage reaction with butylamine in water, the conjugation with a ligand (moenomycin), and experiments that demonstrate that the fully armed probes interact with penicillin-binding protein 1b.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
286338
DL-Isoserine, 98%
C3H7NO3
06054
L-Isoserine, ≥98.0% (TLC)
C3H7NO3