EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
A colorimetric and fluorometric microplate assay for the detection of microcystin-LR in drinking water without preconcentration.
PMID 12176093
Abstract
Protein phosphatase inhibition assays currently used for the detection of cyanobacterial peptide hepatotoxins in drinking water require an enrichment step using C18 cartridges to achieve lower the detection limit. This paper describes a colorimetric and fluorometric protein phosphatase inhibition method for the direct detection of microcystin-LR (MCYST-LR) in drinking water without complex clean-up steps and preconcentration procedures. In this assay three different substrates, p-nitrophenyl phosphate (p-NPP) and two fluorogenic compounds, 4-methylumbelliferyl phosphate (MUP) and 6,8-difluoro-4-methylumbelliferyl phosphate DiFMUP), were tested. The detection limits of the assay are 0.25 and 0.1 microg/l using colorimetric and fluorometric methods, respectively. These levels are well below the provisional guideline value for MCYST-LR of 1 microg/l of drinking water. The detection limit of the fluorometric method is comparable to that of the classical ELISA test. Although both the latter tests allow the detection of MCYST-LR in drinking water directly without pretreatment, the protein phosphatase inhibition assay remain less expensive and therefore more attractive for use in the routine assessment of drinking water contamination by microcystins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8883
4-Methylumbelliferyl phosphate, phosphatase substrate
C10H9O6P