EMAIL THIS PAGE TO A FRIEND
Molecular Biology of the Cell
Clastosome: a subtype of nuclear body enriched in 19S and 20S proteasomes, ubiquitin, and protein substrates of proteasome.
PMID 12181345
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MABE329
Anti-c-Fos Antibody, clone 2G9C3, clone 2G9C3, from mouse