EMAIL THIS PAGE TO A FRIEND
Organic letters
Studies aimed at the total synthesis of azadirachtin. A modeled connection of C-8 and C-14 in azadirachtin.
PMID 12182578
Abstract
[reaction: see text] Studies on the connection between the right and left segments of azadirachtin are described. The Ireland-Claisen rearrangement of Li-enolate of the modeled ester with dichlorodimethylsilane in toluene afforded the desired limonoid framework stereoselectively in good yield.