EMAIL THIS PAGE TO A FRIEND
Organic letters
Concise total synthesis of (+/-)-aloperine and 6-epi-aloperine.
PMID 12182590
Abstract
[reaction: see text] Total synthesis of aloperine and 6-epi-aloperine is reported. The crucial steps of the synthetic strategy are an aza-annulation reaction and an intermolecular Diels-Alder reaction. The synthetic plan proceeds from commercially available piperidine-2-ethanol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
546704
Aloperine, 96%
C15H24N2