EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Hair dye-sensitized hairdressers: the cross-reaction pattern with new generation hair dyes.
PMID 12190619
Abstract
Hair dye allergies are a frequent cause of occupational skin disease among hairdressers. Conventional hair dyes contain 4-phenylenediamine (PPD), 2, 5-diaminotoluene sulfate (DTS) and 2-nitro-4-phenylenediamine (ONPPD) as allergens. In new generation hair dyes, FD & C and D & C dyes are used in the hair dye formulations. This study investigated the cross-reaction pattern of new generation hair dyes among hairdressers (n = 40) with a known allergy to PPD and/or DTS and/or ONPPD. In the 40 hairdressers no positive reactions were observed to the single FD & C and D & C dyes. In two hairdressers, doubtful reactions were observed to one or more of the hair dye formulations. The data from this study suggest that for hairdressers sensitized to PPD and/or DTS and/or ONPPD this new generation of hair dyes is a safe alternative for use in their salons.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
154334
2,5-Diaminotoluene sulfate, 97%
C7H10N2 · H2SO4
N21200
2-Nitro-1,4-phenylenediamine, 95%
C6H7N3O2