EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V
Polyvinylalcohol substituted with triethyleneglycolmonoethylether as a new material for preparation of solid dispersions of hydrophobic drugs.
PMID 12191696
Abstract
Among the different methods used to increase the aqueous drug solubility, the preparation of a solid dispersion with a soluble carrier represents an interesting formulative approach. We substituted polyvinylalcohol with triethyleneglycolmonoethylether and obtained a suitable material for the formulation of a solid dispersion of progesterone, by spray-drying. In particular, we evaluated the influence of the polyvinylalcohol substitution degree and the polymer-drug weight ratios in the preparative mixture on the progesterone dissolution rate in the aqueous environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
568554
Tri(ethylene glycol) monoethyl ether, technical grade
C8H18O4