EMAIL THIS PAGE TO A FRIEND
Clinical Chemistry
Simultaneous analysis of nicotine, nicotine metabolites, and tobacco alkaloids in serum or urine by tandem mass spectrometry, with clinically relevant metabolic profiles.
PMID 12194923