EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Organic modifiers for the separation of organic acids and bases by liquid chromatography.
PMID 12198859
Abstract
A straight-chain alcohol or diol additive in the mobile phase was used to modify and improve the HPLC separation of organic acids and bases. Incorporation of 2% 1-butanol, 1% 1,2-hexanediol, or 0.25% 1,2-octanediol into an aqueous mobile phase greatly improved the separation of alkane carboxylic acids on a silica C18 column, both in terms of separation time and peak shape. When 1.5% 1-hexanol, 0.09% 1-decanol or 0.01% 1-dodecanol was added to an acetonitrile-water (30:70) mobile phase, much sharper peaks and better resolution were obtained for aromatic bases separated on an underivatized polystyrene-divinylbenzene column. The mobile phase additive is believed to coat the stationary phase surface by a dynamic equilibrium. The coated surface is more hydrophilic and facilitates the efficient partitioning of analytes between the mobile and stationary phases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
213705
1,2-Octanediol, 98%
C8H18O2