EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Jasmine absolute (Jasminum grandiflora L.) and its mode of action on guinea-pig ileum in vitro.
PMID 12203263
Abstract
Jasmine (Jasminum grandiflorum L.) is used in aromatherapy as a holistic treatment for apathy, hysteria, uterine disorders and childbirth, muscle relaxation and coughs. Its stimulant nature, on inhalation, has been shown both in animals and man. Jasmine has a spasmolytic activity on guinea-pig ileum and rat uterus in vitro. The mechanism of action of the spasmolytic activity, studied in vitro using a guinea-pig ileum smooth muscle preparation, was postsynaptic and not atropine-like. The spasmolytic effect of jasmine absolute was most likely to be mediated through cAMP, and not through cGMP. The mode of action in vitro resembled that of geranium, lavender and peppermint oils. The contradictory effect in vitro and in vivo is probably due to the solely physiological effects of jasmine absolute in vitro (producing a relaxation) compared with that in vivo, where it has a strong psychological input, producing a stimulant effect in man and enhanced movement in animals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W259802 Jasmin absolute, FG