EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
An umpolung approach to cis-hyponitrite complexes.
PMID 12203596
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
460990
Sodium trans-hyponitrite hydrate
N2Na2O2 · xH2O