EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Determination of isofraxidin in Sarcandra glabra (Thunb.) Nakai by HPLC].
PMID 12205867
Abstract
To establish a method for the determination of isofraxidin in Sarcandra glabra. Using HPLC with mu-Bondapak C18 column, acetonitrile -0.1% phosphoric acid solution(20:80) as mobile phase and detection wavelength at 344 nm. The recovery and RSD were 98.1% and 1.9% (n = 5) respectively. This method is rapid, simple and accurate, thus may be used for the quality control of this drug.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91214
Isofraxidin, analytical standard
C11H10O5