EMAIL THIS PAGE TO A FRIEND
Hua xi yi ke da xue xue bao = Journal of West China University of Medical Sciences = Huaxi yike daxue xuebao
[The synthesis of multifunctional groups and multibranched compounds: IX. Synthesis of several branched building block].
PMID 12205918
Abstract
The branched building plays a pivotal part in the study of Dendrimers. In order to synthesized Dendrimers more effectively, we designed and synthesized four new branched building blocks via 2-4 steps starting from diethyl malonate and pentaerythritol. Their structures were confirmed by IR and 1HNMR etc. They compare favourably with previous branched building blocks as regards to steric hindrance and branched effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
246549
Diethylmalonic acid, 98%
C7H12O4