EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Enantioselective analysis of ibuprofen in human plasma by anionic cyclodextrin-modified electrokinetic chromatography.
PMID 12207314
Abstract
This paper reports the development of a rapid method for the enantioselective analysis of the nonsteroidal anti-inflammatory drug ibuprofen in human plasma by capillary electrophoresis employing the anionic cyclodextrin-modified electrokinetic chromatography mode. Sample cleanup was carried out by acidification with HCl followed by liquid-liquid extraction with hexane:isopropanol (99:1 v/v). The complete enantioselective analysis was performed within 10 min, using 100 mmol L(-1) phosphoric acid/triethanolamine buffer, pH 2.6, containing 2.0% w/v sulfated beta-cyclodextrin as chiral selector; fenoprofen, another nonsteroidal anti-inflammatory drug, was used as internal standard. The calibration curves were linear over the concentration range of 0.25-125.0 microg mL(-1) for each enantiomer of ibuprofen. The mean recoveries for ibuprofen enantiomers were up to 85%. The enantiomers studied could be quantified at three different concentrations (0.5, 5.0 and 50.0 microg mL(-1)) with a coefficient of variation and relative error not higher than 15%. The quantitation limit was 0.2 microg mL(-1) for (+)-(S)- and (-)-(R)-ibuprofen using 1 mL of human plasma. The plasma endogenous compounds and other drugs did not interfere with the present assay. The analysis of real plasma samples obtained from a healthy volunteer after administration of 600 mg of racemic ibuprofen showed a maximum plasma level of 29.6 and 39.9 microg mL(-1) of (-)-(R)- and (+)-(S)-ibuprofen, respectively, and the area under plasma concentration-time curve AUC(0-infinity) (+)-(S)/AUC(0-infinity) (-)-(R) ratio was 1.87.
Related Materials