EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Preparative separation of isomeric sulfophthalic acids by conventional and pH-zone-refining counter-current chromatography.
PMID 12214685
Abstract
Two modes of high-speed counter-current chromatography (HSCCC) were applied to separate 3- and 4-sulfophthalic acid from a mixture. Conventional HSCCC was useful for the separation of up to several hundred milligram quantities of these positional isomers, while pH-zone-refining CCC was implemented successfully to separations at the multigram level. The conventional HSCCC separations were performed with a standard J-type HSCCC system that has a superior resolution but a lower level of retention of the stationary phase of the biphasic solvent system used (acidified n-butanol-water). The pH-zone-refining CCC separations were performed with an X-type HSCCC system (a cross-axis system) that has a higher capability for retention of the stationary phase. The purified positional isomers (over 99% pure as determined by HPLC) were characterized by 1H NMR and negative ion electrospray ionization mass spectrometry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
292362
4-Sulfophthalic acid trisodium salt, technical grade
C8H3Na3O7S