EMAIL THIS PAGE TO A FRIEND
Waste management (New York, N.Y.)
Adsorption of phenol and m-chlorophenol on organobentonites and repeated thermal regeneration.
PMID 12214970
Abstract
Experimental investigations were conducted on the adsorption characteristics of phenol and m-chlorophenol by organobentonites. The organobentonites were prepared by modifying natural bentonite with various quaternary ammonium salts including tetramethylammonium bromide, hexadecyltrimethylammonium bromide, benzyl-triethylammonium bromide, tetraethylammonium bromide and cetylpyridinium bromide. The adsorption characteristics of phenol and chlorophenol by these organobentonites were examined in detail. The empirical Freundlich isotherm was found to describe well the equilibrium adsorption data. Thermal regeneration of spent organobentonites was also investigated and operating conditions of 200 degrees C and 2 h heating were found to yield very good results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C62808
3-Chlorophenol, 98%
C6H5ClO