EMAIL THIS PAGE TO A FRIEND
American journal of clinical pathology
Langerhans cell histiocytosis immunohistochemical expression of fascin, a dendritic cell marker.
PMID 12219775
Abstract
Langerhans cell histiocytosis (LCH) is a clonal disorder believed to be derivedfrom cells of the dendritic system. Fascin, a 55-kd actin-bundling protein, represents a highly selective marker for dendritic cells of lymphoid tissues and peripheral blood and is involved in the formation of dendritic processes in maturing epidermal Langerhans cells. Since lesional cells of LCH may represent Langerhans cells arrested at an early stage of activation, immunohistochemical expression offascin in epidermal Langerhans cells and in the lesional cells of 34 cases of LCH was evaluated in paraffin sections using an immunoalkaline phosphatase technique. Though epidermal Langerhans cells were nonreactive for fascin, lesional cells in all LCH cases exhibited immunoreactivityforfascin, CD1a, and S-100 protein. Variation in staining intensity was observed in some cases, possibly reflecting differences in cell maturation or activation. Involved tissues included bone, soft tissue, lymph node, thyroid, orbit, and extradural cranial tissue. Immunoreactivity of lesional cells of LCH for fascin supports their derivation from cells of the dendritic system and represents another alteration in the phenotype of Langerhans cells that is associated with maturation, migration, culture, or clonal expansion.