EMAIL THIS PAGE TO A FRIEND
Molecular biology of the cell
Purification and identification of secernin, a novel cytosolic protein that regulates exocytosis in mast cells.
PMID 12221138
Abstract
After permeabilization with the pore-forming toxin streptolysin-O mast cells can be triggered to secrete by addition of both calcium and a GTP analogue. If stimulation is delayed after permeabilization, there is a progressive decrease in the extent of secretion upon stimulation, eventually leading to a complete loss of the secretory response. This loss of secretory response can be retarded by the addition of cytosol from other secretory tissues, demonstrating that the response is dependent on a number of cytosolic proteins. We have used this as the basis of a bioassay to purify Secernin 1, a novel 50-kDa cytosolic protein that appears to be involved in the regulation of exocytosis from peritoneal mast cells. Secernin 1 increases both the extent of secretion and increases the sensitivity of mast cells to stimulation with calcium.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9514 Superose® 12 Prep Grade