EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Bulky trialkylsilyl acetylenes in the Cadiot-Chodkiewicz cross-coupling reaction.
PMID 12227823
Abstract
Bulky trialkylsilyl-protected alkynes such as triethylsilyl (TES), tert-butyldimethylsilyl (TBS), and triisopropylsilyl (TIPS) acetylenes underwent the Cadiot-Chodkiewicz cross-coupling reaction with different bromoalkynes to form a variety of synthetically useful unsymmetrical diynes in good yields. The diyne alcohol 10 was transformed regio- and stereoselectively into enynes by hydrotelluration, carbometalation, and reduction reactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
399264
(tert-Butyldimethylsilyl)acetylene, 99%
C8H16Si
346977
(Triethylsilyl)acetylene, 97%
C8H16Si