EMAIL THIS PAGE TO A FRIEND
Atencion primaria
[Robinin intoxication].
PMID 12237034
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53609
Robinin, ≥90.0% (HPLC)
C33H40O19