EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Intercalation and controlled release of pharmaceutically active compounds from a layered double hydroxide.
PMID 12240066
Abstract
A series of pharmaceutically active compounds including diclofenac, gemfibrozil, ibuprofen, naproxen, 2-propylpentanoic acid, 4-biphenylacetic acid and tolfenamic acid can be reversibly intercalated into a layered double hydroxide, initial studies suggest that these materials may have application as the basis of a novel tuneable drug delivery system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196487
4-Biphenylacetic acid, 98%
C14H12O2